man

Test

CAS RN 126370-67-0

NNH2

CAS RN 126370-67-0

IUPAC Name: 6-phenylpyridin-3-amine
Synonyms: 6-phenylpyridin-3-amine
Molecular Weight (g/mol): 170.22
Molecular Formula: C11H10N2
InChi Key: FLIQYTXJLWGVBG-UHFFFAOYSA-N
SMILES: NC1=CN=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results