man

Test

CAS RN 126533-97-9

ONNSO

CAS RN 126533-97-9

IUPAC Name: 2-(morpholin-4-yl)-1,3-thiazole-4-carbaldehyde
Synonyms: 2-(morpholin-4-yl)-1,3-thiazole-4-carbaldehyde
Molecular Weight (g/mol): 198.24
Molecular Formula: C8H10N2O2S
InChi Key: JPJSUXPYPAQDDL-UHFFFAOYSA-N
SMILES: O=CC1=CSC(=N1)N1CCOCC1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results