man

Test

CAS RN 127-40-2

CH3CH3CH3CH3HOH3CCH3CH3OHCH3H3CH3C

CAS RN 127-40-2

IUPAC Name: (1R)-4-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-18-[(1R,4R)-4-hydroxy-2,6,6-trimethylcyclohex-2-en-1-yl]-3,7,12,16-tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaen-1-yl]-3,5,5-trimethylcyclohex-3-en-1-ol
Synonyms: (1R)-4-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-18-[(1R,4R)-4-hydroxy-2,6,6-trimethylcyclohex-2-en-1-yl]-3,7,12,16-tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaen-1-yl]-3,5,5-trimethylcyclohex-3-en-1-ol trans-lutein all-trans-xanthophyll 3,3'-dihydroxy-α-carotene xanthophylls gamma lutein leutin LUT phylloxanthin bo-xan
Molecular Weight (g/mol): 568.89
Molecular Formula: C40H56O2
InChi Key: KBPHJBAIARWVSC-RGZFRNHPSA-N
SMILES: C\C(\C=C\C=C(/C)\C=C\[C@H]1C(C)=C[C@H](O)CC1(C)C)=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)C[C@@H](O)CC1(C)C

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results