accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 128427-10-1

H3CCH3H3COOHN(S)HOOH

CAS RN 128427-10-1

IUPAC Name: tert-butyl N-[(2S)-1,4-dihydroxybutan-2-yl]carbamate
Synonyms: tert-butyl N-[(2S)-1,4-dihydroxybutan-2-yl]carbamate
Molecular Weight (g/mol): 205.25
Molecular Formula: C9H19NO4
InChi Key: KLRRFBSWOIUAHZ-ZETCQYMHSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CO)CCO

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products