CAS RN 1291-48-1
CAS RN 1291-48-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific Acros 1,1'-Ferrocenedimethanol, 98%, Thermo Scientific Chemicals
CAS: 1291-48-1 Molecular Formula: C12H14FeO2 Molecular Weight (g/mol): 246.09 MDL Number: MFCD02091688 Synonym: ferrocene derivatives,1,1'-bis hydroxymethyl ferrocen,1,1'-ferrocenedimethanol,1,1 inverted exclamation marka-ferrocenedimethanol SMILES: [Fe].OCc1cccc1.OCc1cccc1