CAS RN 129486-91-5
CAS RN 129486-91-5
IUPAC Name:
2-methyl-1H-imidazole-5-carbothioamide
Synonyms:
2-methyl-3H-imidazole-4-carbothioamide
Molecular Weight (g/mol):
141.19
Molecular Formula:
C5H7N3S
InChi Key:
MNYPQSNAWPZXRV-UHFFFAOYSA-N
SMILES:
CC1=NC=C(N1)C(N)=S
1
–
1
of
1
results
1
Thermo Scientific Maybridge 2-Methyl-1H-imidazole-4-carbothioamide, 97%, Thermo Scientific™
CAS: 129486-91-5 Molecular Formula: C5H7N3S Molecular Weight (g/mol): 141.192 MDL Number: MFCD03659724 InChI Key: MNYPQSNAWPZXRV-UHFFFAOYSA-N Synonym: 2-methyl-1h-imidazole-4-carbothioamide,1h-imidazole-5-carbothioamide,2-methyl,1h-imidazole-5-carbothioamide, 2-methyl,acmc-1c27g,amino 2-methylimidazol-4-yl methane-1-thione,2-methyl-3h-imidazole-4-carbothioamide PubChem CID: 2796714 IUPAC Name: 2-methyl-1H-imidazole-5-carbothioamide SMILES: CC1=NC=C(N1)C(=S)N