CAS RN 130-37-0
CAS RN 130-37-0
IUPAC Name:
sodium 2-methyl-1,4-dioxo-1,2,3,4-tetrahydronaphthalene-2-sulfonate
Synonyms:
menadione sodium bisulfite vitamin K3 sodium bisulfite menaphthone sodium bisulphite bisulfito sodico de menadiona menadioni natrii bisulfis sodium menadione bisulfite menadione sodium sulfonate bisulfite sodique de menadione menaphthone sodium bisulfite natrium menadionsulfonicum
Molecular Weight (g/mol):
276.24
Molecular Formula:
C11H9NaO5S
InChi Key:
XDPFHGWVCTXHDX-UHFFFAOYNA-M
SMILES:
[Na+].CC1(CC(=O)C2=CC=CC=C2C1=O)S([O-])(=O)=O
1
–
1
of
1
results
1
Medchem Express Menadione bisulfite sodium, MedChemExpress
MedChemExpress Menadione bisulfite (sodium) is used as an agent to induce acute oxidative stress, and to function as a plant-defense activator against several pathogens.
