CAS RN 1301706-46-6
CAS RN 1301706-46-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 1-Benzyloxycarbonyl-(2S,4R)-2-cyano-4-fluoropyrrolidine, 97%, Thermo Scientific™
CAS: 1301706-46-6 Molecular Formula: C13H13FN2O2 MDL Number: MFCD08458685 Synonym: n-cbz-trans-4-fluoro-l-prolinonitrile,1-benzyloxycarbonyl-2s,4r-2-cyano-4-fluoropyrrolidine,benzyl 2s,4r-2-cyano-4-fluoropyrrolidine-1-carboxylate,n-benzyloxycarbonyl-trans-4-fluoro-l-prolinonitrile,2s,4r-benzyl 2-cyano-4-fluoropyrrolidine-1-carboxylate,benzyl 2s,4r-2-cyano-4-fluoro-pyrrolidine-1-carboxylate