man

Test

CAS RN 13021-40-4

KOHOHOOOO

CAS RN 13021-40-4

IUPAC Name: dihydroxycyclohex-5-ene-1,2,3,4-tetrone potassium
Synonyms: rhodizonic acid potassium
Molecular Weight (g/mol): 209.17
Molecular Formula: C6H2KO6
InChi Key: UGFHFZSZCJFOTO-UHFFFAOYSA-N
SMILES: [K].OC1=C(O)C(=O)C(=O)C(=O)C1=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results