man

Test

CAS RN 1305-79-9

CaOO

CAS RN 1305-79-9

IUPAC Name: calcium dioxidanediide
Synonyms: calcium peroxide
Molecular Weight (g/mol): 72.08
Molecular Formula: CaO2
InChi Key: LHJQIRIGXXHNLA-UHFFFAOYSA-N
SMILES: [Ca++].[O-][O-]
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results