man

Test

CAS RN 13125-66-1

H3CMgBr

CAS RN 13125-66-1

IUPAC Name: magnesium(2+) heptan-1-ide bromide
Synonyms: magnesium(2+) heptan-1-ide bromide
Molecular Weight (g/mol): 203.41
Molecular Formula: C7H15BrMg
InChi Key: GRYDGXUVWLGHPL-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].CCCCCC[CH2-]

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results