accessibility menu, dialog, popup

UserName

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 131379-14-1

CNZnBr

CAS RN 131379-14-1

IUPAC Name: zinc(2+) 4-cyanobenzen-1-ide bromide
Synonyms: zinc(2+) 4-cyanobenzen-1-ide bromide
Molecular Weight (g/mol): 247.4
Molecular Formula: C7H4BrNZn
InChi Key: HEOMKTNNZDZMJZ-UHFFFAOYSA-M
SMILES: [Zn++].[Br-].N#CC1=CC=[C-]C=C1
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products