accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 131379-16-3

H3COOCZnI

CAS RN 131379-16-3

IUPAC Name: 4-(ethoxycarbonyl)benzen-1-ide; iodozincylium
Synonyms: 4-(ethoxycarbonyl)benzen-1-ide; iodozincylium
Molecular Weight (g/mol): 341.45
Molecular Formula: C9H9IO2Zn
InChi Key: DVAYVJGYZRYJEV-UHFFFAOYSA-M
SMILES: [Zn+]I.CCOC(=O)C1=CC=[C-]C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products