accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13139-16-7

CH3(S)(S)HNOOCH3H3CCH3OOHH3C

CAS RN 13139-16-7

IUPAC Name: (2S,3R)-2-{[(tert-butoxy)carbonyl]amino}-3-methylpentanoate
Synonyms: (2S,3R)-2-[(tert-butoxycarbonyl)amino]-3-methylpentanoate
Molecular Weight (g/mol): 230.29
Molecular Formula: C11H20NO4
InChi Key: QJCNLJWUIOIMMF-SFYZADRCSA-M
SMILES: CC[C@@H](C)[C@H](NC(=O)OC(C)(C)C)C([O-])=O
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products