accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1314-96-1

SrS

CAS RN 1314-96-1

IUPAC Name: sulfanylidenestrontium
Synonyms: strontium sulfide
Molecular Weight (g/mol): 119.68
Molecular Formula: SSr
InChi Key: XXCMBPUMZXRBTN-UHFFFAOYSA-N
SMILES: S=[Sr]

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products