CAS RN 1315-06-6
CAS RN 1315-06-6
IUPAC Name:
stannaneselone
Synonyms:
tin selenide
Molecular Weight (g/mol):
199.7
Molecular Formula:
H2SeSn
InChi Key:
RTZUOCZUTSLBGC-UHFFFAOYSA-N
SMILES:
[Se]=[SnH2]
1
–
1
of
1
results
1
Tin selenide, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 1315-06-6 Molecular Formula: SnSe MDL Number: MFCD00054049