accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1315339-86-6

SH2NFFFCl

CAS RN 1315339-86-6

IUPAC Name: 4-chloro-3-(trifluoromethyl)benzene-1-carbothioamide
Synonyms: 4-chloro-3-(trifluoromethyl)benzenecarbothioamide
Molecular Weight (g/mol): 239.64
Molecular Formula: C8H5ClF3NS
InChi Key: MPBUEWPKJPREPS-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC(=C(Cl)C=C1)C(F)(F)F
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products