accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 131980-30-8

CAS RN 131980-30-8

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(3-cyanophenyl)propanoate
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(3-cyanophenyl)propanoate
Molecular Weight (g/mol): 289.31
Molecular Formula: C15H17N2O4
InChi Key: FDQDHMZKOPOWFE-LBPRGKRZSA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC(=C1)C#N)C([O-])=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products