CAS RN 132-27-4
CAS RN 132-27-4
IUPAC Name:
sodium [1,1'-biphenyl]-2-olate
Synonyms:
sodium ortho-phenylphenate
Molecular Weight (g/mol):
192.19
Molecular Formula:
C12H9NaO
InChi Key:
KSQXVLVXUFHGJQ-UHFFFAOYSA-M
SMILES:
[Na+].[O-]C1=CC=CC=C1C1=CC=CC=C1