CAS RN 13235-36-4
CAS RN 13235-36-4
IUPAC Name:
tetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate tetrahydrate
Synonyms:
tetrasodium ion(4-) edta tetrahydrate
Molecular Weight (g/mol):
452.23
Molecular Formula:
C10H20N2Na4O12
InChi Key:
XFLNVMPCPRLYBE-UHFFFAOYSA-J
SMILES:
O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O