man

Test

CAS RN 13254-34-7

CH3H3CCH3CH3HO

CAS RN 13254-34-7

IUPAC Name: 2,6-dimethylheptan-2-ol
Synonyms: 2-heptanol, 2,6-dimethyl-
Molecular Weight (g/mol): 144.26
Molecular Formula: C9H20O
InChi Key: HGDVHRITTGWMJK-UHFFFAOYSA-N
SMILES: CC(C)CCCC(C)(C)O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results