CAS RN 133-08-4
CAS RN 133-08-4
IUPAC Name:
1,3-diethyl 2-butylpropanedioate
Synonyms:
1,3-diethyl 2-butylpropanedioate
Molecular Weight (g/mol):
216.28
Molecular Formula:
C11H20O4
InChi Key:
RPNFNBGRHCUORR-UHFFFAOYSA-N
SMILES:
CCCCC(C(=O)OCC)C(=O)OCC
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar Diethyl n-butylmalonate, 99%, Thermo Scientific™
CAS: 133-08-4 Molecular Formula: C11H20O4 Molecular Weight (g/mol): 216.28 MDL Number: MFCD00009170 InChI Key: RPNFNBGRHCUORR-UHFFFAOYSA-N Synonym: diethyl butylmalonate,diethyl n-butylmalonate,butylmalonic acid diethyl ester,diethyl 2-butylmalonate,ethyl butylmalonate,propanedioic acid, butyl-, diethyl ester,unii-cyy2q744wb,cyy2q744wb,n-butylmalonic acid diethyl ester,malonic acid, butyl-, diethyl ester PubChem CID: 67244 IUPAC Name: diethyl 2-butylpropanedioate SMILES: CCCCC(C(=O)OCC)C(=O)OCC