CAS RN 133261-33-3
CAS RN 133261-33-3
IUPAC Name:
4-(trifluoromethyl)cyclohexane-1-carboxylic acid
Synonyms:
4-(trifluoromethyl)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol):
196.17
Molecular Formula:
C8H11F3O2
InChi Key:
LMEAZIIFLVDISW-UHFFFAOYSA-N
SMILES:
OC(=O)C1CCC(CC1)C(F)(F)F
1
–
1
of
1
results