man

Test

CAS RN 13363-51-4

H2NSNH2S

CAS RN 13363-51-4

IUPAC Name: benzene-1,4-dicarbothioamide
Synonyms: benzene-1,4-dicarbothioamide
Molecular Weight (g/mol): 196.29
Molecular Formula: C8H8N2S2
InChi Key: USHPIZCRGQUHGN-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=C(C=C1)C(N)=S

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results