accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13372-81-1

OHN(E)(E)

CAS RN 13372-81-1

IUPAC Name: (E)-N-[(2E)-3-phenylprop-2-en-1-ylidene]hydroxylamine
Synonyms: (E)-N-[(2E)-3-phenylprop-2-en-1-ylidene]hydroxylamine
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: RUQDOYIAKHIMAN-DAAQNPAKSA-N
SMILES: O\N=C\C=C\C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products