man

Test

CAS RN 1338-43-8

CH3OOOHOOHOH

CAS RN 1338-43-8

IUPAC Name: 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl (9Z)-octadec-9-enoate
Synonyms: sorbitan oleate
Molecular Weight (g/mol): 428.61
Molecular Formula: C24H44O6
InChi Key: NWGKJDSIEKMTRX-NZAQQJATSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results