accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13393-64-1

CH2CH2(E)

CAS RN 13393-64-1

IUPAC Name: (5E)-deca-1,5,9-triene
Synonyms: (5E)-deca-1,5,9-triene
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: GMGYODAMPOELNZ-MDZDMXLPSA-N
SMILES: C=CCC\C=C\CCC=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products