CAS RN 134-91-8
CAS RN 134-91-8
IUPAC Namn:
bis[4-(diethylamino)phenyl]methanol
Synonymer:
bis[4-(diethylamino)phenyl]methanol
Molekylvikt (g/mol):
326.48
Molekylär sammansättning:
C21H30N2O
InChi Key:
WCIQBUVXZZYFJP-UHFFFAOYSA-N
SMILES:
CCN(CC)C1=CC=C(C=C1)C(O)C1=CC=C(C=C1)N(CC)CC
1
–
3
av
3
Resultat
1
bis(4-Diethylaminophenyl)methanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
bis(4-Diethylaminophenyl)methanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
bis(4-Diethylaminophenyl)methanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.