accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13450-88-9

GaBrBrBr

CAS RN 13450-88-9

IUPAC Name: gallium(3+) tribromide
Synonyms: gallium(III) bromide
Molecular Weight (g/mol): 309.44
Molecular Formula: Br3Ga
InChi Key: SRVXDMYFQIODQI-UHFFFAOYSA-K
SMILES: [Ga+3].[Br-].[Br-].[Br-]
Physical Form 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products