CAS RN 13466-78-9
CAS RN 13466-78-9
IUPAC Name:
3,7,7-trimethylbicyclo[4.1.0]hept-3-ene
Synonyms:
monoterpenes 3-carene, (R)- delta(sup 3)-carene (r)-3-carene 3-carene, (s)-(cis)-isomer carene 3-carene, (r)- 3-carene (S)-(cis)-isomer 3-carene 3-carene, (r)-isomer
Molecular Weight (g/mol):
136.24
Molecular Formula:
C10H16
InChi Key:
BQOFWKZOCNGFEC-UHFFFAOYNA-N
SMILES:
CC1=CCC2C(C1)C2(C)C
1
–
1
of
1
results
1
3-Carene, 90%, stabilized, Thermo Scientific Chemicals
CAS: 13466-78-9 Molecular Formula: C10H16 Molecular Weight (g/mol): 136.23 MDL Number: MFCD00001315 InChI Key: BQOFWKZOCNGFEC-UHFFFAOYSA-N Synonym: 3-carene,carene,3,7,7-trimethylbicyclo 4.1.0 hept-3-ene,delta-3-carene,monoterpenes,alpha-carene,bicyclo 4.1.0 hept-3-ene, 3,7,7-trimethyl,+--delta3-carene,+ car-3-ene,+--3-carene PubChem CID: 26049 ChEBI: CHEBI:35661 IUPAC Name: 4,7,7-trimethylbicyclo[4.1.0]hept-3-ene SMILES: CC1=CCC2C(C1)C2(C)C