man

Test

CAS RN 13472-30-5

NaNaNaNaOSiOOO

CAS RN 13472-30-5

IUPAC Name: tetrasodium silicate
Synonyms: sodium orthosilicate
Molecular Weight (g/mol): 184.04
Molecular Formula: Na4O4Si
InChi Key: POWFTOSLLWLEBN-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[Na+].[Na+].[O-][Si]([O-])([O-])[O-]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results