accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1347815-41-1

NINH2N

CAS RN 1347815-41-1

IUPAC Name: 2-amino-5-iodopyridine-3-carbonitrile
Synonyms: 2-amino-5-iodopyridine-3-carbonitrile
Molecular Weight (g/mol): 245.02
Molecular Formula: C6H4IN3
InChi Key: NJLCSLOABAUGHW-UHFFFAOYSA-N
SMILES: NC1=NC=C(I)C=C1C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products