CAS RN 135065-76-8
CAS RN 135065-76-8
IUPAC Name:
tert-butyl 2-(hydroxymethyl)morpholine-4-carboxylate
Synonyms:
tert-butyl 2-(hydroxymethyl)morpholine-4-carboxylate
Molecular Weight (g/mol):
217.27
Molecular Formula:
C10H19NO4
InChi Key:
FJYBLMJHXRWDAQ-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)N1CCOC(CO)C1