CAS RN 1350702-25-8
CAS RN 1350702-25-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar Azido-PEG3-carboxytetramethylrhodamine 110 conjugate, Thermo Scientific Chemicals
CAS: 1350702-25-8 Molecular Formula: C33H38N6O7 MDL Number: MFCD22380734 Synonym: Azide-Fluor 545