accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 135159-25-0

H3COOCH3BOHHOOH3C

CAS RN 135159-25-0

IUPAC Name: (2,4,6-trimethoxyphenyl)boronic acid
Synonyms: 2,4,6-trimethoxyphenylboronic acid
Molecular Weight (g/mol): 212.01
Molecular Formula: C9H13BO5
InChi Key: PKLRXZVPEQJTTJ-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(B(O)O)C(OC)=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products