accessibility menu, dialog, popup

UserName

CAS RN 1351761-44-8

CH3HNNNFFFNHFONOOCH3

CAS RN 1351761-44-8

IUPAC Name: N4-ethyl-N2-[5-fluoro-2-methoxy-4-(morpholine-4-carbonyl)phenyl]-5-(trifluoromethyl)pyrimidine-2,4-diamine
Synonyms: N4-ethyl-N2-[5-fluoro-2-methoxy-4-(morpholine-4-carbonyl)phenyl]-5-(trifluoromethyl)pyrimidine-2,4-diamine
Molecular Weight (g/mol): 443.4
Molecular Formula: C19H21F4N5O3
InChi Key: XCFLWTZSJYBCPF-UHFFFAOYSA-N
SMILES: CCNC1=NC(NC2=C(OC)C=C(C(=O)N3CCOCC3)C(F)=C2)=NC=C1C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results