accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 135296-80-9

H2OH2OH2OClOsClCl

CAS RN 135296-80-9

IUPAC Name: trichloroosmium trihydrate
Synonyms: trichloroosmium trihydrate
Molecular Weight (g/mol): 350.63
Molecular Formula: Cl3H6O3Os
InChi Key: UACQLNPCDXDCID-UHFFFAOYSA-K
SMILES: O.O.O.Cl[Os](Cl)Cl
Grade 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products