man

Test

CAS RN 13530-65-9

ZnOCrOOO

CAS RN 13530-65-9

IUPAC Name: zinc(2+) dioxochromiumbis(olate)
Synonyms: zinc chromate
Molecular Weight (g/mol): 181.37
Molecular Formula: CrO4Zn
InChi Key: NDKWCCLKSWNDBG-UHFFFAOYSA-N
SMILES: [Zn++].[O-][Cr]([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results