CAS RN 1354039-86-3
CAS RN 1354039-86-3
IUPAC Name:
2-{1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-yl}-2-phenyl-N-(propan-2-yl)acetamide
Synonyms:
2-{1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl}-N-isopropyl-2-phenylacetamide
Molecular Weight (g/mol):
518.65
Molecular Formula:
C31H38N2O5
InChi Key:
HXHOBPVRRPCTLG-UHFFFAOYNA-N
SMILES:
COC1=C(OC)C=C(CC2N(CCC3=CC(OC)=C(OC)C=C23)C(C(=O)NC(C)C)C2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
Tocris Bioscience™ ACT 335827

CAS: 1354039-86-3 Molecular Formula: C31H38N2O5 Molecular Weight (g/mol): 518.654