CAS RN 135579-85-0
CAS RN 135579-85-0
IUPAC Name:
3-cyanopropan-1-ide; bromozincylium
Synonyms:
3-cyanopropan-1-ide; bromozincylium
Molecular Weight (g/mol):
213.38
Molecular Formula:
C4H6BrNZn
InChi Key:
HEHCQTGEYNYOSF-UHFFFAOYSA-M
SMILES:
[Zn+]Br.[CH2-]CCC#N
1
–
1
of
1
results