accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13574-13-5

CH3H3CCH3OOHN(S)OOOO

CAS RN 13574-13-5

IUPAC Name: (2S)-5-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoate
Synonyms: (2S)-5-(benzyloxy)-2-[(tert-butoxycarbonyl)amino]-5-oxopentanoate
Molecular Weight (g/mol): 336.37
Molecular Formula: C17H22NO6
InChi Key: AJDUMMXHVCMISJ-ZDUSSCGKSA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CCC(=O)OCC1=CC=CC=C1)C([O-])=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products