CAS RN 1364488-67-4
CAS RN 1364488-67-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ YH 239-EE
Promotes survival of muscle stem cells (MuSCs)
2
Medchem Express YH239-EE, MedChemExpress
MedChemExpress YH239-EE, ethyl ester of the free carboxylic acid compound YH239, is a potent p53-MDM2 antagonizing and apoptosis-inducing agent.
