CAS RN 13650-49-2
CAS RN 13650-49-2
IUPAC Name:
2-amino-5-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Synonyms:
2-amino-5-[(tert-butoxycarbonyl)amino]pentanoic acid
Molecular Weight (g/mol):
232.28
Molecular Formula:
C10H20N2O4
InChi Key:
GLZZMUULAVZVTA-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NCCCC(N)C(O)=O
1
–
1
of
1
results
1
Ndelta-Boc-L-ornithine, 98%, Thermo Scientific Chemicals
CAS: 13650-49-2 Molecular Formula: C10H20N2O4 Molecular Weight (g/mol): 232.28 MDL Number: MFCD00037219 InChI Key: GLZZMUULAVZVTA-ZETCQYMHSA-N Synonym: h-orn boc-oh,s-2-amino-5-tert-butoxycarbonylamino-pentanoic acid,boc-l-orn-oh,boc-l-orn-oh,ndelta-boc-l-ornithine,ndelta-boc-l-ornithine,a-boc-l-ornithine,l-ornithine, n5-1,1-dimethylethoxy carbonyl,l-ornithine, n5-1,1-dimethylethoxy carbonyl,n∼5∼-tert-butoxycarbonyl-l-ornithine,n∼5∼-tert-butoxycarbonyl-l-ornithine PubChem CID: 6993436 IUPAC Name: (2S)-2-amino-5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid SMILES: CC(C)(C)OC(=O)NCCCC(C(=O)O)N