accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13676-54-5

OONNOO

CAS RN 13676-54-5

IUPAC Name: 1-(4-{[4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)phenyl]methyl}phenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(4-{[4-(2,5-dioxopyrrol-1-yl)phenyl]methyl}phenyl)pyrrole-2,5-dione
Molecular Weight (g/mol): 358.35
Molecular Formula: C21H14N2O4
InChi Key: XQUPVDVFXZDTLT-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1C1=CC=C(CC2=CC=C(C=C2)N2C(=O)C=CC2=O)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products