man

Test

CAS RN 13682-61-6

ClClClClKAu

CAS RN 13682-61-6

IUPAC Name: gold(3+) potassium tetrachloride
Synonyms: gold(3+) potassium tetrachloride
Molecular Weight (g/mol): 377.86
Molecular Formula: AuCl4K
InChi Key: JMONCQYBAMKVQQ-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[K+].[Au+3]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results