accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13707-88-5

HClH3CCH3NHOHOCH2

CAS RN 13707-88-5

IUPAC Name: hydrogen 1-[2-(prop-2-en-1-yl)phenoxy]-3-[(propan-2-yl)amino]propan-2-ol chloride
Synonyms: alprenolol hydrochloride alprenolol hcl
Molecular Weight (g/mol): 285.81
Molecular Formula: C15H24ClNO2
InChi Key: RRCPAXJDDNWJBI-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(C)NCC(O)COC1=CC=CC=C1CC=C
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products