CAS RN 137071-78-4
CAS RN 137071-78-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress 3,6-Dichlorotrimellitic acid
MedChemExpress 3,6-Dichlorotrimellitic acid is the key precursor that is used for preparing a variety of dichlorinated fluoresceins and rhodamines such as TET and HEX.
