accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13734-38-8

H3CCH3H3CONHOOCH3CH3H3COHO

CAS RN 13734-38-8

IUPAC Name: 3-(tert-butoxy)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: 3-(tert-butoxy)-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 261.32
Molecular Formula: C12H23NO5
InChi Key: BPYLRGKEIUPMRJ-UHFFFAOYNA-N
SMILES: CC(C)(C)OCC(NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products