man

Test

CAS RN 138124-32-0

CH3H3CCH3NHNHOCH3H3CCH3CH3CH3H3COH3CH3CCH3ClMn

CAS RN 138124-32-0

IUPAC Name: 2,4-di-tert-butyl-6-{[(2-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}cyclohexyl)amino]methylidene}cyclohexa-2,4-dien-1-one manganese chloride
Synonyms: 2,4-di-tert-butyl-6-{[(2-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}cyclohexyl)amino]methylidene}cyclohexa-2,4-dien-1-one manganese chloride
Molecular Weight (g/mol): 637.23
Molecular Formula: C36H54ClMnN2O2
InChi Key: YRVXCOIDMFNGIJ-UHFFFAOYNA-M
SMILES: [Cl-].[Mn].CC(C)(C)C1=CC(=CNC2CCCCC2NC=C2C=C(C=C(C2=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results