man

Test

CAS RN 13813-23-5

IIIPr

CAS RN 13813-23-5

IUPAC Name: praseodymium(3+) triiodide
Synonyms: praseodymium(3+) triiodide
Molecular Weight (g/mol): 521.62
Molecular Formula: I3Pr
InChi Key: PVEVRIVGNKNWML-UHFFFAOYSA-K
SMILES: [I-].[I-].[I-].[Pr+3]
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results